Learn More
Biomatik Corporation Human TBP2 (Thioredoxin Binding Protein 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61036
Encompass_Preferred
Human TBP2 (Thioredoxin Binding Protein 2) ELISA Kit for detecting TBP2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q9H3M7. Target Synonyms: TXNIP; HHCPA78; THIF; VDUP1; Thioredoxin Interacting Protein; Vitamin D3-Upregulated Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0368