Learn More
Biomatik Corporation Human UST2 (Urotensin 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61149
Encompass_Preferred
Human UST2 (Urotensin 2) ELISA Kit for detecting UST2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: O95399. Target Synonyms: UII; U-II; UCN2; PRO1068; Urotensin-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0442