Learn More
Biomatik Corporation Human tPA (Plasminogen Activator, Tissue) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61172
Encompass_Preferred
Human tPA (Tissue-type Plasminogen Activator) ELISA Kit for detecting tPA in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.63-40ng/ml. Sensitivity: 0.38ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P00750. Target Synonyms: PLAT; T-PA; TPA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0457