Learn More
Biomatik Corporation Human VWF (Von Willebrand Factor) ELISA Kit, 96 tests
Catalog No. 501500473
Encompass_Preferred
Human VWF (Von Willebrand Factor) ELISA Kit for detecting VWF in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.94ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P04275. Target Synonyms: F8VWF; VWD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0473 Supplier Biomatik Corporation Supplier No. EKE61195
Human VWF (Von Willebrand Factor) ELISA Kit for detecting VWF in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.94ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P04275. Target Synonyms: F8VWF; VWD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.