Learn More
Biomatik Corporation Human SRD5a1 (Steroid 5 Alpha Reductase 1) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKE61347
Human SRD5a1 (Steroid 5 Alpha Reductase 1) ELISA Kit for detecting SRD5a1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P18405. Restrictions: For Research Use Only. Not for use in diagnostic procedures.