Learn More
Biomatik Corporation Human RTN4 (Reticulon 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61351
Encompass_Preferred
Human RTN4 (Reticulon 4) ELISA Kit for detecting RTN4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q9NQC3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0575