Learn More
Biomatik Corporation Human ALPL (Alkaline Phosphatase, Liver/Bone/Kidney) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61360
Encompass_Preferred
The Human ALPL (Alkaline Phosphatase, Liver/Bone/Kidney) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 78.13-5000 pg/mL. Sensitivity: 46.88 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P05186. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0581