Learn More
Biomatik Corporation Human AD (Adropin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61365
Encompass_Preferred
The Human AD (Adropin) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 12.5-800 pg/mL. Sensitivity: 7.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: Q6UWT2. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0584