Learn More
Biomatik Corporation Human GPX1 (Glutathione Peroxidase 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61376
Encompass_Preferred
The Human GPX1 (Glutathione Peroxidase 1) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P07203. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0591