Learn More
Biomatik Corporation Human TACSTD2 (Tumor-associated calcium signal transducer 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61386
Encompass_Preferred
Human TACSTD2 (Tumor-associated Calcium Signal Transducer 2) ELISA Kit for detecting TACSTD2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500pg/ml. Sensitivity: 4.69pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P09758. Target Synonyms: EGP-1; EGP1; GA733-1; GA7331; GP50; M1S1; TROP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0599