Learn More
Biomatik Corporation Human MR-ProADM (Mid-regional pro-adrenomedullin) ELISA Kit , 96 tests
Supplier:  Biomatik Corporation EKE61390
Encompass_Preferred
The Human MR-ProADM (Mid-regional pro-adrenomedullin) ELISA Kit  allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 15.63-1000 pg/mL. Sensitivity: 9.38 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Catalog No. 50-150-0601