Learn More
Biomatik Corporation HumanAng1-7 (Angiotensin1-7) ELISAKit, 96 tests
Supplier:  Biomatik Corporation EKE61420
Encompass_Preferred
Human Ang1-7 (Angiotensin 1-7) ELISA Kit for detecting Ang1-7 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P01019. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0619