Learn More
Biomatik Corporation HumanpNF-H (PhosphorylatedNeurofilamentH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61429
Encompass_Preferred
Human pNF-H (Phosphorylated Neurofilament H) ELISA Kit for detecting pNF-H in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P12036. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0622