Learn More
Biomatik Corporation Human sRANKL (Soluble Receptor Activator of Nuclear factor-kB Ligand) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61441
Encompass_Preferred
Human sRANKL (Soluble Receptor Activator of Nuclear factor-kB Ligand) ELISA Kit for detecting sRANKL in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: O14788 (140~317aa). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0628