Learn More
Biomatik Corporation Mouse NT-proBNP (N-terminal pro-Brain Natriuretic Peptide) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61729
Encompass_Preferred
Mouse NT-proBNP (N-terminal pro-Brain Natriuretic Peptide) ELISA Kit for detecting NT-proBNP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P40753. Target Synonyms: NT-Pro-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0646