Learn More
Biomatik Corporation Human ANG2 (Angiopoietin-2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57010
Encompass_Preferred
The Human ANG2 (Angiopoietin-2) ELISA Kit allows for quantitative determination of ANG2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 46.875-3000 pg/mL. Sensitivity: 28.125 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q9UID3. Restriction: For Research Use Only.
Catalog No. 50-150-0657