Learn More
Biomatik Corporation Human CRP (C-Reactive Protein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57020
Encompass_Preferred
The Human CRP (C-Reactive Protein) ELISA Kit allows for quantitative determination of CRP in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 31.2-2000 pg/mL. Sensitivity: 2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P02741. Restriction: For Research Use Only.
Catalog No. 50-150-0666