Learn More
Biomatik Corporation Human CXCL10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57021
Encompass_Preferred
The Human CXCL10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit allows for quantitative determination of CXCL10 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 7.813-500 pg/mL. Sensitivity: 4.688 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P02778. Restriction: For Research Use Only.
Catalog No. 50-150-0667