Learn More
Biomatik Corporation Human CXCL13 (C-X-C motif chemokine 13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57022
Encompass_Preferred
The Human CXCL13 (C-X-C motif chemokine 13) ELISA Kit allows for quantitative determination of CXCL13 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: O43927. Restriction: For Research Use Only.
Catalog No. 50-150-0668