Learn More
Biomatik Corporation Human CYR61 (Cysteine Rich Protein, Angiogenic Inducer 61) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57025
Encompass_Preferred
The Human CYR61 (Cysteine Rich Protein, Angiogenic Inducer 61) ELISA Kit allows for quantitative determination of CYR61 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: O00622. Restriction: For Research Use Only.
Catalog No. 50-150-0671