Learn More
Biomatik Corporation Human CCL11 (Eotaxin) ELISA Kit, 96 tests
Catalog No. 501500678
Encompass_Preferred
The Human CCL11 (Eotaxin) ELISA Kit allows for quantitative determination of CCL11 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P51671. Restriction: For Research Use Only.
Catalog No. 50-150-0678 Supplier Biomatik Corporation Supplier No. EKF57032
Add to Cart
Add to Cart
The Human CCL11 (Eotaxin) ELISA Kit allows for quantitative determination of CCL11 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P51671. Restriction: For Research Use Only.