Learn More
Biomatik Corporation Human SELE (E-Selectin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57034
Encompass_Preferred
Human SELE (E-Selectin) ELISA Kit for detecting SELE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16581. Target Synonyms: E-Selectin; SELE; CD62E; CD62E; ELAM1; LECAM2; SELE; CD62 antigen-like family member E; CD62E; CD62E antigen; ELAM; ELAM-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0680