Learn More
Biomatik Corporation Human FN1 (Fibronectin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57038
Encompass_Preferred
The Human FN1 (Fibronectin) ELISA Kit allows for quantitative determination of FN1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P02751. Restriction: For Research Use Only.
Catalog No. 50-150-0684