Learn More
Biomatik Corporation Human GDF15 (Growth Differentiation Factor 15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57044
Encompass_Preferred
The Human GDF15 (Growth Differentiation Factor 15) ELISA Kit allows for quantitative determination of GDF15 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 23.438-1500 pg/mL. Sensitivity: 14.063 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q99988. Restriction: For Research Use Only.
Catalog No. 50-150-0690