Learn More
Biomatik Corporation Human GH (Growth Hormone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57046
Encompass_Preferred
The Human GH (Growth Hormone) ELISA Kit allows for quantitative determination of GH in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 25-1600 pg/mL. Sensitivity: 15 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01241. Restriction: For Research Use Only.
Catalog No. 50-150-0692