Learn More
Biomatik Corporation Human HBEGF (Proheparin-binding EGF-like growth factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57049
Encompass_Preferred
The Human HBEGF (Proheparin-binding EGF-like growth factor) ELISA Kit allows for quantitative determination of HBEGF in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 31.2-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q99075. Restriction: For Research Use Only.
Catalog No. 50-150-0695