Learn More
Biomatik Corporation Human sICAM-1/CD54 (Soluble Intercellular Adhesion Molecule 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57051
Encompass_Preferred
Human sICAM-1/CD54 (Soluble Intercellular Adhesion Molecule 1) ELISA Kit for detecting sICAM-1; CD54 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05362. Target Synonyms: sICAM-1; CD54; ICAM1; BB2; P3.58. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0697