Learn More
Biomatik Corporation Human IGF-1 (Insulin-Like Growth Factor 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57053
Encompass_Preferred
The Human IGF-1 (Insulin-Like Growth Factor 1) ELISA Kit allows for quantitative determination of IGF-1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05019. Restriction: For Research Use Only.
Catalog No. 50-150-0699