Learn More
Biomatik Corporation Human IL1RL1 (Interleukin-1 receptor-like 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57066
Encompass_Preferred
The Human IL1RL1 (Interleukin-1 receptor-like 1) ELISA Kit allows for quantitative determination of IL1RL1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5000 pg/mL. Sensitivity: 46.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q01638. Restriction: For Research Use Only.
Catalog No. 50-150-0712