Learn More
Biomatik Corporation Human IL-4 (Interleukin 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57071
Encompass_Preferred
The Human IL-4 (Interleukin 4) ELISA Kit allows for quantitative determination of IL-4 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05112. Restriction: For Research Use Only.
Catalog No. 50-150-0717