Learn More
Biomatik Corporation Human IL-6 (Interleukin 6) ELISA Kit, 96 tests
Catalog No. 501500719
Encompass_Preferred
The Human IL-6 (Interleukin 6) ELISA Kit allows for quantitative determination of IL-6 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 4.69-300 pg/mL. Sensitivity: 2.813 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05231. Restriction: For Research Use Only.
Catalog No. 50-150-0719 Supplier Biomatik Corporation Supplier No. EKF57073
The Human IL-6 (Interleukin 6) ELISA Kit allows for quantitative determination of IL-6 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 4.69-300 pg/mL. Sensitivity: 2.813 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05231. Restriction: For Research Use Only.