Learn More
Biomatik Corporation Human SERPINA4 (Kallistatin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57077
Encompass_Preferred
Human SERPINA4 (Kallistatin) ELISA Kit for detecting SERPINA4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29622. Target Synonyms: SERPINA4; Kallistatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0723