Learn More
Biomatik Corporation Human LEP (Leptin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57079
Encompass_Preferred
The Human LEP (Leptin) ELISA Kit allows for quantitative determination of LEP in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 31.2-2000 pg/mL. Sensitivity: 18.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P41159. Restriction: For Research Use Only.
Catalog No. 50-150-0725