Learn More
Biomatik Corporation Human LUM (Lumican) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57081
Encompass_Preferred
The Human LUM (Lumican) ELISA Kit allows for quantitative determination of LUM in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P51884. Restriction: For Research Use Only.
Catalog No. 50-150-0727