Learn More
Biomatik Corporation Human MDC (Macrophage Derived Chemokine) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57083
Encompass_Preferred
The Human MDC (Macrophage Derived Chemokine) ELISA Kit allows for quantitative determination of MDC in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: O75078. Restriction: For Research Use Only.
Catalog No. 50-150-0729