Learn More
Biomatik Corporation Human C-ERC (Mesothelin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57084
Encompass_Preferred
The Human C-ERC (Mesothelin) ELISA Kit allows for quantitative determination of C-ERC in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0730