Learn More
Biomatik Corporation Human MMP-1 (Matrix Metalloproteinase 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57086
Encompass_Preferred
The Human MMP-1 (Matrix Metalloproteinase 1) ELISA Kit allows for quantitative determination of MMP-1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 46.875 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P03956. Restriction: For Research Use Only.
Catalog No. 50-150-0732