Learn More
Biomatik Corporation Human NGF/NGFβ (Beta-nerve growth factor) ELISA Kit, 96 tests
Catalog No. 501500738
Encompass_Preferred
The Human NGF/NGFβ (Beta-nerve growth factor) ELISA Kit allows for quantitative determination of NGF/NGFβ in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01138. Restriction: For Research Use Only.
Catalog No. 50-150-0738 Supplier Biomatik Corporation Supplier No. EKF57092
The Human NGF/NGFβ (Beta-nerve growth factor) ELISA Kit allows for quantitative determination of NGF/NGFβ in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01138. Restriction: For Research Use Only.