Learn More
Biomatik Corporation Human OPN (Osteopontin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57095
Encompass_Preferred
The Human OPN (Osteopontin) ELISA Kit allows for quantitative determination of OPN in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 156-10000 pg/mL. Sensitivity: 93.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P10451. Restriction: For Research Use Only.
Catalog No. 50-150-0741