Learn More
Biomatik Corporation Human RBP4 (Retinol-binding protein 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57104
Encompass_Preferred
The Human RBP4 (Retinol-binding protein 4) ELISA Kit allows for quantitative determination of RBP4 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P02753. Restriction: For Research Use Only.
Catalog No. 50-150-0750