Learn More
Biomatik Corporation Human RETN (Resistin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57107
Encompass_Preferred
Human RETN (Resistin) ELISA Kit for detecting RETN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9HD89. Target Synonyms: Resistin; RETN; ADSF; Adipose tissue-specific secretory factor; HXCP1; RETN1; RSTNXCP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0753