Learn More
Biomatik Corporation Human CLEC3B (Tetranectin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57110
Encompass_Preferred
The Human CLEC3B (Tetranectin) ELISA Kit allows for quantitative determination of CLEC3B in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50 ng/mL. Sensitivity: 0.469 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05452. Restriction: For Research Use Only.
Catalog No. 50-150-0756