Learn More
Biomatik Corporation Human TIMP-1 (Metalloproteinase inhibitor 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57116
Encompass_Preferred
Human TIMP-1 (Metalloproteinase inhibitor 1) ELISA Kit for detecting TIMP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01033. Target Synonyms: TIMP-1; TIMP1; CLGI; EPA; EPO; HCI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0762