Learn More
Biomatik Corporation Human TNFRSF4/OX40 (Tumor necrosis factor receptor superfamily member 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57122
Encompass_Preferred
Human TNFRSF4/OX40 (Tumor necrosis factor receptor superfamily member 4) ELISA Kit for detecting TNFRSF4; OX40 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43489. Target Synonyms: OX40; TNFRSF4; CD134; TAX transcriptionally-activated glycoprotein 1 receptor; ACT35 antigen; OX40L receptor; ACT-135; CD134 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; OX40lymphoid activation antigene ACT35; tumor necrosis factor receptor superfamily member 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0768