Learn More
Biomatik Corporation Human TNFSF4/OX40L (Tumor necrosis factor ligand superfamily member 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57124
Encompass_Preferred
Human TNFSF4/OX40L (Tumor necrosis factor ligand superfamily member 4) ELISA Kit for detecting TNFSF4; OX40L in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23510. Target Synonyms: OX40L; TNFSF4; CD252; gp34; CD134 ligand; CD134L; CD252 antigen; Glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX-40L; OX4OL; TXGP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0770