Learn More
Biomatik Corporation Human VCAM-1 (Vascular cell adhesion protein 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57126
Encompass_Preferred
Human VCAM-1 (Vascular cell adhesion protein 1) ELISA Kit for detecting VCAM-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19320. Target Synonyms: VCAM-1; CD106; sVCAM-1; CD106 antigen; INCAM-100; L1CAM; vascular cell adhesion molecule 1; vascular cell adhesion protein 1; V-CAM 1; VCAM1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0772