Learn More
Biomatik Corporation Human VEGF (Vascular Endothelial Cell Growth Factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57127
Encompass_Preferred
Human VEGF (Vascular Endothelial Cell Growth Factor) ELISA Kit for detecting VEGF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15692. Target Synonyms: VEGF; VAS; Vasculotropin; VEGFA; VPF; MVCD1; vascular endothelial growth factor A; Vascular permeability factor; VEGF-A; VPFvascular endothelial growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0773