Learn More
Biomatik Corporation Human FABP3 (Fatty Acid Binding Protein 3, Muscle and Heart) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57131
Encompass_Preferred
The Human FABP3 (Fatty Acid Binding Protein 3, Muscle and Heart) ELISA Kit allows for quantitative determination of FABP3 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P05413. Restriction: For Research Use Only.
Catalog No. 50-150-0777