Learn More
Biomatik Corporation Human CTNI (Cardiac Troponin-I) ELISA Kit, 96 tests
Catalog No. 501500779
Encompass_Preferred
The Human CTNI (Cardiac Troponin-I) ELISA Kit allows for quantitative determination of cTnI in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 12.5-800 pg/mL. Sensitivity: 2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0779 Supplier Biomatik Corporation Supplier No. EKF57133
The Human CTNI (Cardiac Troponin-I) ELISA Kit allows for quantitative determination of cTnI in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 12.5-800 pg/mL. Sensitivity: 2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.