Learn More
Biomatik Corporation Human TF (Transferrin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57146
Encompass_Preferred
Human TF (Transferrin) ELISA Kit for detecting TF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02787. Target Synonyms: TF; Transferrin; PRO1557; PRO2086; TFQTL1; Siderophilin; Beta-1 metal-binding globulin; Beta-1 metal-binding globulin; serotransferrin; Serotransferrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0792